Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Trefoil Factor 3 Monoclonal Antibody (5UCLNT3), Biotin, eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. 13475882
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-475-882 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-475-882 Supplier Invitrogen™ Supplier No. 13475882
  • $353.00
    $264.65 / Each of 1

    Save $88.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

The MAb 5UCLNT3 recognizes mouse and rat Trefoil Factor 3. Trefoil Factor 3 (TFF3) is a member of the trefoil family, which are characterized by having at least one copy of the trefoil motif, a 40-amino acid domain that contains three conserved disulfides. TFF3 is a small peptide hormone secreted by mucus-producing cells and by epithelial cells of multiple tissues, including the gastrointestinal tract, kidney, and breast. TFF3 plays an essential role in both mucosal surface maintenance and restitution. TFF3 facilitates restoration of the intestinal epithelium as a protective barrier against injury by inhibiting apoptosis and promoting survival and migration of epithelial cells. TFF3 also plays a role in inducing airway epithelial ciliated cell differentiation. Levels of TFF3 change after renal injury and urinary levels can be used as a biomarker for kidney tubular injury. TFF3 may also play a role in breast tumor progression and tumor cell dissemination.

Trefoil peptides are protease resistant molecules secreted throughout the gut that play a role in mucosal healing. Trefoil peptides contain three intrachain disulfide bonds, forming the trefoil motif, or P-domain and clustered in human chromosome 21q22.3. Trefoil Factor 3 (TFF3) is one of three trefoil peptides secreted by epithelial cells that line mucus membranes in the large and small intestines and lower respiratory tract. TFF3 is mainly active as a disulfide-linked homodimer and may have a role in promoting cell migration in healing processes. It is involved in the maintenance and repair of the intestinal mucosa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Trefoil Factor 3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 5UCLNT3
Concentration 0.5 mg/mL
Conjugate Biotin
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene TFF3
Gene Accession No. Q03191, Q62395
Gene Alias hITF; hP1.B; Intestinal trefoil factor; ITF; mITF; P1B; Polypeptide P1.B; rITF; rP1.B; Tff3; TFI; Trefoil factor 3; trefoil factor 3 (intestinal); trefoil factor 3, intestinal
Gene Symbols TFF3
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21786, 25563
Target Species Mouse, Rat
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.