Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ TREML1/TLT-1 Antibody Pair
SDP

Catalog No. NB397946 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
15 Plates
5 Plates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB397946 15 Plates
NB397945 5 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB397946 Supplier Novus Biologicals™ Supplier No. NBP31165915PLATES
Add to Cart
Add to Cart

Antibody Pair

specifically detects in Human samples. It is validated for ELISA.

Specifications

Assay Range 28.13 to 1800 pg/mL (example only, lot dependent)
Assay Sensitivity 28.13 pg/mL (example only; lot dependent)
Conjugate Unconjugated
Detection Method Sandwich ELISA
Immunoassay Kit Format Sandwich ELISA Kit
Product Type Sandwich ELISA
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 340205
Gene Symbol TREML1
Quantity 15 Plates
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym dJ238O23.3, GLTL1825, PRO3438, TLT-1, TLT1MGC119173, trem-like transcript 1 protein, triggering receptor expressed on myeloid cells-like 1, Triggering receptor expressed on myeloid cells-like protein 1
Target Human
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.