Learn More
Description
Peptide Sequence: MGPGARRQGR IVQGRPEMCF CPPPTPLPPL RILTLGTHTP TPCSSPGSAA GTYPTMGSQA LPPGPMQTLI FFDMEATGLP FSQPKVTELC LLAVHRCALE.
Specifications
Specifications
| Antigen | TREX1 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 1B1 |
| Concentration | 0.34 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | TREX1 |
| Gene Accession No. | Q9NSU2 |
| Gene Alias | 1661; 3' repair exonuclease 1; 3'-5' exonuclease TREX1; AGS1; AU041952; CRV; Deoxyribonuclease III; DNase III; DRN3; HERNS; RGD1309596; three prime exonuclease 1; three prime repair exonuclease 1; three-prime repair exonuclease 1; Trex1; trophoblast expressed 1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
