Learn More
Invitrogen™ TREX2 Polyclonal Antibody

Description
Human TREX2 shares 88.6% amino acid (aa) sequence identity with mouse TREX2. Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: HELA whole cell, MCF-7 whole cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
Specifications
Specifications
| Antigen | TREX2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 500 μg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5mg BSA and 0.05mg sodium azide |
| Gene | TREX2 |
| Gene Accession No. | Q9BQ50 |
| Gene Alias | 3'-5' exonuclease TREX2; 3'-5' exonuclease TREX2 long form; RGD1562245; Three prime repair exonuclease 2; Trex2 |
| Gene Symbols | TREX2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.