Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRF-2 Antibody [mFluor Violet 450 SE], R&D Systems™
SDP

Catalog No. NB641004
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB641004 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB641004 Supplier R&D Systems Supplier No. AF5635MFV450

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Goat Polyclonal Antibody

TRF-2 Polyclonal specifically detects TRF-2 in Human samples. It is validated for Western Blot.

Specifications

Antigen TRF-2
Applications Western Blot
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Formulation 50mM Sodium Borate
Gene Alias Telomeric DNA-binding protein, telomeric repeat binding factor 2, Telomeric repeat binding protein 2, telomeric repeat-binding factor 2, TERF2, TRBF2, TRF2, TTAGGG repeat binding factor 2, TTAGGG repeat-binding factor 2
Host Species Goat
Immunogen E. coli-derived recombinant human TRF2, Gln78-Thr238, Accession # Q15554
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 7014
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.