Learn More
Description
Suggested positive control: antigen standard for TERF2 (transient overexpression lysate), Daudi, HeLa, HL-60, or Jurkat. Immunogen sequence homology: rat (86%).
Specifications
Specifications
| Antigen | TRF2 |
| Applications | ChIP Assay, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% sodium azide |
| Gene | TERF2 |
| Gene Accession No. | Q15554 |
| Gene Alias | Telomeric DNA-binding protein; telomeric repeat binding factor 2; telomeric repeat binding protein 2; telomeric repeat-binding factor 2; TERF2; TRBF2; TRF2; TTAGGG repeat-binding factor 2 |
| Gene Symbols | TERF2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
