Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRF2 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501558468
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-155-8468 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-155-8468 Supplier Bethyl Laboratories, Inc Supplier No. A300796AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

TRF2 is a ubiquitously expressed protein that is implicated in the control of telomere length (1). TRF2, like TRF1 contains a Myb-related DNA binding motif. It binds to duplex TTAGGG repeats and is localized to all human telomeres in metaphase chromosomes (1). TRF2 is thought to protect chromosome ends by maintaining the correct structure at telomere termini (2). The use of mutant forms of TRF2 has implicated a role for TRF2 in the prevention of senescence in primary human cells (2). Recently, it has been shown that inhibition of TRF2 resulted in apoptosis in a subset of mammalian cell types (3).

Specifications

Antigen TRF2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene TERF2
Gene Accession No. Q15554
Gene Alias TRBF2, TRF2
Gene Symbols TERF2
Host Species Rabbit
Immunogen Between 375 and 425
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7014
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.