Learn More
Invitrogen™ TRF4-1 Polyclonal Antibody

Description
This antibody detects various isoforms of TRF4-1. Suggested positive control: Hela whole cell extract, HeLa cell lysate. Immunogen sequence: GTASLKAVHH MSSPAI.
Specifications
Specifications
| Antigen | TRF4-1 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.5% BSA and 0.02% sodium azide |
| Gene | TENT4A |
| Gene Accession No. | Q5XG87 |
| Gene Alias | DNA polymerase kappa; DNA polymerase sigma; LAK1; LAK-1; non-canonical poly(A) RNA polymerase PAPD7; PAP associated domain containing 7; PAP-associated domain-containing protein 7; Papd7; POLK; POLS; polymerase (DNA directed) sigma; polymerase (DNA-directed) sigma; Tent4a; Terminal guanylyltransferase; Terminal nucleotidyltransferase 4A; Terminal uridylyltransferase 5; topoisomerase-related function protein 4-1; TRAMP-like complex polyadenylate polymerase; TRF4; TRF41; TRF4-1; TUTase 5; TUTASE5 |
| Gene Symbols | TENT4A |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.