Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRF4-1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA129801
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA129801 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA129801 Supplier Invitrogen™ Supplier No. PA129801
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detects a band ~92 kDa.

TRF4-1 is a nuclear protein whose expression is cell cycle-regulated at a post-transcriptional level. Two genes, TRF4-1 and TRF4-2, belonging to this family have been identified. TRF4 is also required to maintain a different aspect of higher order chromosome structure, sister chromatid cohesion.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRF4-1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene TENT4A
Gene Accession No. Q5XG87
Gene Alias DNA polymerase kappa; DNA polymerase sigma; LAK1; LAK-1; non-canonical poly(A) RNA polymerase PAPD7; PAP associated domain containing 7; PAP-associated domain-containing protein 7; Papd7; POLK; POLS; polymerase (DNA directed) sigma; polymerase (DNA-directed) sigma; Tent4a; Terminal guanylyltransferase; Terminal nucleotidyltransferase 4A; Terminal uridylyltransferase 5; topoisomerase-related function protein 4-1; TRAMP-like complex polyadenylate polymerase; TRF4; TRF41; TRF4-1; TUTase 5; TUTASE5
Gene Symbols TENT4A
Host Species Rabbit
Immunogen Synthetic peptide containing amino acids 447-462 (GTASLKAVHHMSSPAI) of human TRF4-1.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11044
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.