Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRFP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA579658
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA579658 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA579658 Supplier Invitrogen™ Supplier No. PA579658
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: rat brain tissue, human PANC-1 whole cell.

MED20 is a subunit of the Mediator complex, a multiprotein coactivator of RNA transcription that interacts with DNA-bound transcriptional activators, RNA polymerase II, and general initiation factors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRFP
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide
Gene MED20
Gene Accession No. Q5XIE9, Q9H944, Q9R0X0
Gene Alias 1110011O05Rik; 2410115I17Rik; AU018348; hTRFP; Med20; Mediator complex subunit 20; mediator of RNA polymerase II transcription subunit 20; PRO0213; Trf (TATA binding protein-related factor)-proximal homolog; Trf (TATA binding protein-related factor)-proximal protein homolog; TRFP; TRF-proximal protein homolog; ubiquitin specific protease 49; Usp49
Gene Symbols MED20
Host Species Rabbit
Immunogen E. coli-derived human TRFP / MED20 recombinant protein (Position: M1-R212).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 316209, 56771, 9477
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.