Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRHR Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01675297
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-675-297 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-675-297 Supplier Invitrogen™ Supplier No. PA141469
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TRHR Polyclonal Antibody for Western Blot, IHC (P)

TRH-R is a 398 amino acid integral membrane protein with six transmembrane domains and belongs to G-protein coupled receptor 1 family. It acts as a receptor for the signaling hormone TRH and activates the inositol phospholipid-calcium-protein kinase C transduction pathway upon binding with TRH. Reports suggest that TRH-R is a neurotransmitter and neuro regulator that controls dimerization of the TRH receptor and potentiates hormone-induced receptor trafficking. It is known to widely express in the thyrotroph cells of the anterior pituitary.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRHR
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% gelatin and 0.05% sodium azide
Gene TRHR
Gene Accession No. P34981
Gene Alias MGC141920; Thyroliberin receptor; thyrotropin releasing hormone receptor; thyrotropin-releasing hormone receptor; TRH receptor; TRH1 Receptor; Trhr; TRH-R; TRH-R1
Gene Symbols TRHR
Host Species Rabbit
Immunogen A portion of amino acids 350-400 of human TRH-R protein.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 482007, 7201
Target Species Human, Canine, Monkey
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.