Learn More
Medchemexpress LLC HY-125006 5mg Medchemexpress, TRi-1 CAS:246020-68-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1250065MG
Encompass_Preferred
Medchemexpress, HY-125006 5mg TRi-1 CAS:246020-68-8 TRi-1 is a potent, specific and irreversible inhibitor of cytosolic thioredoxin reductase 1 (TXNRD1) , with an IC 50 of 12 nM. TRi-1 has little mitochondrial toxicity for anticancer therapy [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2599
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.