Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tri(4-morpholinyl)phosphine oxide, 99%

Catalog No. AAL1631514
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1631514 25 g
AAL1631506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1631514 Supplier Thermo Scientific Chemicals Supplier No. L1631514
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 4441-12-7 | C24H42 | 330.60 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4441-12-7
MDL Number MFCD00047406

Specifications

Melting Point 188°C to 192°C
Molecular Formula C12H24N3O4P
Quantity 25 g
Beilstein 280155
Synonym trimorpholinophosphine oxide, 4,4',4-phosphoryltrimorpholine, tri 4-morpholinyl phosphine oxide, phosphoric acid trimorpholide, 4-bis morpholin-4-yl phosphoroso morpholine, tmpo;tris-morpholino-phosphine oxide, phosphoric trimorpholide, 5-n-propylbenzoic acid, acmc-2097fl, ksc493m7l
InChI Key WXMQHPKQCPCDQO-UHFFFAOYSA-N
SMILES C1COCCN1P(=O)(N2CCOCC2)N3CCOCC3
IUPAC Name 4-dimorpholin-4-ylphosphorylmorpholine
Molecular Weight (g/mol) 305.32
PubChem CID 78180
Formula Weight 305.32
Percent Purity 99%
Chemical Name or Material Tri(4-morpholinyl)phosphine oxide
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 224-662-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.