Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

β-Tri-Calcium phosphate, ≥98% β to phase basis, Solstice

Catalog No. 60047010 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
10 g
100 g
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-047-010 100 g Poly Bottle
60-047-011 10 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-047-010 Supplier Solstice Supplier No. 49963100G
Add to Cart
Add to Cart

Puriss. p.a.,≥98% β-phase basis (sintered Powder)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-87-4
Molecular Formula Ca3O8P2
Molecular Weight (g/mol) 310.174
MDL Number MFCD00015984
InChI Key QORWJWZARLRLPR-UHFFFAOYSA-H
Synonym calcium phosphate, synthos, tricalcium phosphate, tricalcium diphosphate, calcigenol simple, calcium orthophosphate, beta-tcp, tribasic calcium phosphate, bonarka, calcium phosphate, tribasic
PubChem CID 24456
ChEBI CHEBI:9679
IUPAC Name tricalcium;diphosphate
SMILES [O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[Ca+2].[Ca+2].[Ca+2]

Specifications

Melting Point 1670°C
Color White
Physical Form Solid
Quantity 100 g
UN Number NONH for all modes of transport
Formula Weight 310.18g/mol
Percent Purity ≥98% β to phase basis (sintered Powder)
Packaging Poly Bottle
Density 3.27g/cm3 at 20.5°C
Chemical Name or Material β-tri-Calcium phosphate
RTECSNumber 231-840-8
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.