Learn More
Tri-n-butyltin chloride, 96%
Description
Tri-n-butyltin chloride is used as an endocrine disruptor as well as an inhibitor for the V-ATPases. It is also used as a rodent-repellent for cable coatings. Further, it is used in hot end glass coating. It also employed as a corrosion remover.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Tri-n-butyltin chloride is used as an endocrine disruptor as well as an inhibitor for the V-ATPases. It is also used as a rodent-repellent for cable coatings. Further, it is used in hot end glass coating. It also employed as a corrosion remover.
Solubility
Miscible with alcohol, heptane, benzene and toluene. Immiscible with water.
Notes
Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | -19°C |
| Density | 1.2 |
| Boiling Point | 171°C to 173°C (25 mmHg) |
| Flash Point | 120°C (248°F) |
| Refractive Index | 1.491 |
| Quantity | 1000 g |
| UN Number | UN2788 |
| Beilstein | 3535715 |
| Solubility Information | Miscible with alcohol,heptane,benzene and toluene. Immiscible with water. |
| Formula Weight | 325.51 |
| Show More |
Safety and Handling
| Hazard Category | H301-H312-H315-H319-H360FD-H372 |
| Hazard Statement | GHS H Statement H301-H372-H312-H315-H319 Toxic if swallowed. Causes damage to organs through prolonged or repeated exposure. Harmful in contact with skin. Causes skin irritation. Causes serious eye irritation. |
| Precautionary Statement | P201-P202-P260-P264b-P270-P271-P280i-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P332+P313-P362-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S. |
| EINECSNumber | 215-958-7 |
| RTECSNumber | WH6820000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.