Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tri-n-butyltin methoxide, 97%

Catalog No. AAA133550B
Change view
Click to view available options
Quantity:
100 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA133550B 1000 g
AAA1335522 100 g
AAA1335530 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA133550B Supplier Thermo Scientific Chemicals Supplier No. A133550B
Add to Cart

CAS: 1067-52-3 | C13H30OSn | 321.092 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1067-52-3
Molecular Formula C13H30OSn
Molecular Weight (g/mol) 321.092
MDL Number MFCD00009419
InChI Key KJGLZJQPMKQFIK-UHFFFAOYSA-N
Synonym tributyltin methoxide, stannane, tributylmethoxy, unii-9cff2afn2v, tri-n-butyltin methanolate, 9cff2afn2v, methoxytributyltin, tin, tributylmethoxy, tributyltin methanolate, stannane,tributylmethoxy, stannane, methoxytributyl
PubChem CID 16683411
IUPAC Name tributyl(methoxy)stannane
SMILES CCCC[Sn](CCCC)(CCCC)OC

Specifications

Density 1.12
Boiling Point 124°C to 126°C (4 mmHg)
Flash Point 98°C (208°F)
Refractive Index 1.474
Quantity 1000 g
UN Number UN2788
Beilstein 3600960
Sensitivity Moisture sensitive
Formula Weight 321.09
Percent Purity 97%
Chemical Name or Material Tri-n-butyltin methoxide
Show More Show Less
Hazard Category H301-H312-H315-H319-H372
Hazard Statement GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P260-P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 213-933-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.