Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tri-n-octylphosphine oxide, 98%

Catalog No. aaa1106214
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1106214 25 g
AAA1106222 100 g
AAA1106236 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1106214 Supplier Thermo Scientific Chemicals Supplier No. A1106214
Only null left
Add to Cart

CAS: 78-50-2 | C24H51OP | 386.645 g/mol

Tri-n-octylphosphine oxide is a solvent used in the extraction of metals, especially uranium, zirconium and hafnium. It is also used as an auxiliary reagent in the solvent extraction of metals. It is also used to extract hydrogen bonding organic compounds. Further, it is useful as a capping ligand for the production of quantum dots, such as those consisting of cadmium selenide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tri-n-octylphosphine oxide is a solvent used in the extraction of metals, especially uranium, zirconium and hafnium. It is also used as an auxiliary reagent in the solvent extraction of metals. It is also used to extract hydrogen bonding organic compounds. Further, it is useful as a capping ligand for the production of quantum dots, such as those consisting of cadmium selenide.

Solubility
Soluble in chloroform, toluene and hexane. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-50-2
Molecular Formula C24H51OP
Molecular Weight (g/mol) 386.645
MDL Number MFCD00002083
InChI Key ZMBHCYHQLYEYDV-UHFFFAOYSA-N
Synonym trioctylphosphine oxide, tri-n-octylphosphine oxide, topo, phosphine oxide, trioctyl, cyanex 921, hostarex px 324, trioctyl phosphine oxide, trioctylphosphane oxide, trioctylphosphino-1-one, acmc-209peg
PubChem CID 65577
IUPAC Name 1-dioctylphosphoryloctane
SMILES CCCCCCCCP(=O)(CCCCCCCC)CCCCCCCC

Specifications

Melting Point 52°C to 57°C
Density 0.88
Boiling Point 212°C to 213°C (0.3 mmHg)
Flash Point 252°C (486°F)
Odor Odorless
Quantity 25 g
Beilstein 1796648
Solubility Information Soluble in chloroform,toluene and hexane. Insoluble in water.
Formula Weight 386.65
Percent Purity 98%
Chemical Name or Material Tri-n-octylphosphine oxide
Show More Show Less
Hazard Category H315-H317-H361
Hazard Statement GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P201-P202-P261-P264b-P272-P280-P281-P302+P352-P305+P351+P338-P308+P313-P333+P313-P362-P501c
EINECSNumber 201-121-3
RTECSNumber SZ1662500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.