Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tri(o-tolyl)phosphine 97.0+%, TCI America™
SDP

Catalog No. T102425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T102425G 25 g
T10245G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T102425G Supplier TCI America Supplier No. T102425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6163-58-2
Molecular Formula C21H21P
Molecular Weight (g/mol) 304.373
MDL Number MFCD00008514
InChI Key COIOYMYWGDAQPM-UHFFFAOYSA-N
Synonym tri-o-tolylphosphine, tri o-tolyl phosphine, tris 2-methylphenyl phosphine, tris 2-methylphenyl phosphane, phosphine, tris 2-methylphenyl, tris o-tolyl phosphine, tri-ortho-toylphosphine, tri-ortho-tolylphosphine, phosphine, tri-o-tolyl, tri 2-methylphenyl phosphine
PubChem CID 80271
IUPAC Name tris(2-methylphenyl)phosphane
SMILES CC1=CC=CC=C1P(C2=CC=CC=C2C)C3=CC=CC=C3C

Specifications

Melting Point 124°C
Color White-Yellow
Quantity 25 g
Formula Weight 304.37
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Tri(o-tolyl)phosphine
EINECSNumber (3)-4357
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.