Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Tri-Sil TP (TMSI:Pyridine) Reagent

Catalog No. PI49231 Shop All Thermo Scientific Products
Encompass
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI49231 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI49231 Supplier Thermo Scientific™ Supplier No. TS49231
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Derivatize hydroxyls compounds, particularly carbohydrates with Thermo Scientific™ Tri-Sil TP (TMSI:Pyridine) Reagent.

Derivatize hydroxyls compounds, particularly carbohydrates with Thermo Scientific™ Tri-Sil TP (TMSI:Pyridine) Reagent. Comprising TMSI in pyridine, this reagent silylates alcohols and phenols, organic acids, hydroxylamines, amino acids, carbohydrates, flavonoids, glycols and polyglycols, nucleotides, steroids, hydroxyl acids, barbituates, narcotics, indoles, and vitamins.

Additonal Product Information

Does not react with amines

May be used in the presence of water as long as there is enough reagent present to react with both the water and the sample

TRUSTED_SUSTAINABILITY

Specifications

For Use With GC
Hazards Flammable
Storage 2°C to 8°C
Quantity 25 mL
Standard Components Pyridine, 1-(Trimethylsilyl)-1 H-imidazole
Standard Concentration 65 - 80, 20 - 25%

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.