Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tri-tert-butylphosphonium tetrafluoroborate, 97%

Catalog No. AAL1975206
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1975206 5 g
AAL1975203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1975206 Supplier Thermo Scientific Chemicals Supplier No. L1975206
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 131274-22-1 | C12H28BF4P | 290.13 g/mol

Tri-tert-butylphosphonium tetrafluoroborate is a ligand used in the palladium-catalyzed enantioselective alfa-arylation of N-boc-pyrrolidine. It is also used with a palladium(0)-15-membered, triolefinic, macrocyle in Suzuki cross-coupling reactions of aryl bromides and chlorides. Further, it is used in the Heck coupling of vinyl tosylates with olefins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tri-tert-butylphosphonium tetrafluoroborate is a ligand used in the palladium-catalyzed enantioselective alfa-arylation of N-boc-pyrrolidine. It is also used with a palladium(0)-15-membered, triolefinic, macrocyle in Suzuki cross-coupling reactions of aryl bromides and chlorides. Further, it is used in the Heck coupling of vinyl tosylates with olefins.

Solubility
Soluble in methylene chloride and chloroform. Slightly soluble in terahydro furan. Insoluble in hexane, toluene and water.

Notes
Incompatible with strong oxidizing agents and halogens.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 131274-22-1
Molecular Formula C12H28BF4P
Molecular Weight (g/mol) 290.13
MDL Number MFCD04039975
InChI Key YTJUCJAUJCXFTN-UHFFFAOYSA-O
Synonym tri-tert-butylphosphonium tetrafluoroborate, t-bu 3ph bf4, tri-t-butylphosphonium tetrafluoroborate, fluoroboric acid tri-tert-butylphosphine adduct, tri-tert-butylphosphanium tetrafluoroborate, tbu3p hbf4, ttbp hbf4, t-bu3p hbf4, tbu 3p hbf4, acmc-20a4f9
PubChem CID 2734635
IUPAC Name tritert-butylphosphanium;tetrafluoroborate
SMILES F[B-](F)(F)F.CC(C)(C)[PH+](C(C)(C)C)C(C)(C)C

Specifications

Melting Point 266°C to 267°C
Quantity 5 g
Beilstein 8813613
Sensitivity Hygroscopic
Solubility Information Soluble in methylene chloride and chloroform. Slightly soluble in terahydro furan. Insoluble in hexane,toluene and water.
Formula Weight 290.13
Percent Purity 97%
Chemical Name or Material Tri-tert-butylphosphonium tetrafluoroborate
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.