Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triacetin, CP, Spectrum™ Chemical
SDP

Catalog No. 18614501 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 mL
4 L
Packaging:
Steel Can
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-614-501 500 mL Steel Can
18-614-500 4 L Steel Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-614-501 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. T1098500MLGL

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 102-76-1

Specifications

CAS Max % 1
Boiling Point 258°C
Packaging Steel Can
Quantity 500 mL
Grade Chemically Pure
Physical Form Liquid
Chemical Name or Material Triacetin

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.