Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triacetin, FCC, 98.5%, Spectrum™ Chemical
SDP

Catalog No. 18607502 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
4 L
6 x 500 mL
20 L
4 x 4 L
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-502 4 L Amber Glass Bottle
18-607-501 6 x 500 mL Amber Glass Bottle
18-607-503 4 x 4 L Amber Glass Bottle
18-607-504 20 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-607-502 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. T12004LTGL
Add to Cart
Add to Cart

T1200, 102-76-1, C9H14O6

Spectrum™ Chemical Triacetin, FCC is widely used as humectant, emlusifier, binder in food production. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage, nutritional supplements and personal care applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 102-76-1
Molecular Formula C9H14O6
Molecular Weight (g/mol) 218.21
InChI Key URAYPUMNDPQOKB-UHFFFAOYSA-N
IUPAC Name 1,3-bis(acetyloxy)propan-2-yl acetate
SMILES CC(=O)OCC(COC(C)=O)OC(C)=O

Specifications

Acidity To pass test
CAS Max % 1
Boiling Point 258°C
Packaging Amber Glass Bottle
Refractive Index 1.429-1.431
Quantity 4 L
Percent Purity 98.5%
Grade FCC
Physical Form Liquid
Chemical Name or Material Triacetin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.