Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triacetin (Glycerol Triacetate), Reagents

Catalog No. 01333453
Change view
Click to view available options
Quantity:
120 mL
500 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-333-453 1 L
01-333-454 120 mL
01-333-452 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-333-453 Supplier Reagents Holdings Llc Supplier No. CT1002001C
Only null left
Add to Cart
Add to Cart

Triacetin (Glycerol Triacetate), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 102-76-1
Molecular Formula C9H14O6
Molecular Weight (g/mol) 218.21
InChI Key URAYPUMNDPQOKB-UHFFFAOYSA-N
Synonym 1,2,3-Propanetriol, Glycerol Triacetate
IUPAC Name 1,3-bis(acetyloxy)propan-2-yl acetate
SMILES CC(=O)OCC(COC(C)=O)OC(C)=O

Specifications

Boiling Range 258°C to 259°C
Color Colorless
Boiling Point 258°C
Packaging Glass Bottle
Quantity 1 L
Specific Gravity 1.156
Grade Reagent
Physical Form Liquid
Chemical Name or Material Triacetin (Glycerol Triacetate)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.