Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triacetin, USP, 97-102%, Spectrum™ Chemical
SDP

Catalog No. 18607506 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
4 L
6 x 500 mL
4 x 4 L
200 L
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-506 6 x 500 mL Amber Glass Bottle
18-607-507 4 L Amber Glass Bottle
18-607-508 4 x 4 L Amber Glass Bottle
18-607-510 200 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-607-506 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. TR106500MLGLCS6
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

TR106, 102-76-1, C9H14O6

Spectrum™ Chemical Triacetin, USP is used as an antifungal agent. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 102-76-1
Molecular Formula C9H14O6
Molecular Weight (g/mol) 218.21
InChI Key URAYPUMNDPQOKB-UHFFFAOYSA-N
IUPAC Name 1,3-bis(acetyloxy)propan-2-yl acetate
SMILES CC(=O)OCC(COC(C)=O)OC(C)=O

Specifications

CAS Max % 1
Boiling Point 258°C
Packaging Amber Glass Bottle
Quantity 6 x 500 mL
Percent Purity 97 to 102%
Grade USP
Physical Form Liquid
Chemical Name or Material Triacetin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.