Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triacontane 98.0+%, TCI America™
SDP

Catalog No. T05945G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T05945G 5 g
T059425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T05945G Supplier TCI America Supplier No. T05945G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 638-68-6
Molecular Formula C30H62
Molecular Weight (g/mol) 422.826
MDL Number MFCD00009410
InChI Key JXTPJDDICSTXJX-UHFFFAOYSA-N
Synonym n-triacontane, triacontane, analytical standard, triacontane, acmc-1b4uf, n-triacontane 250mg, ch3-ch2 28-ch3
PubChem CID 12535
ChEBI CHEBI:31006
IUPAC Name triacontane
SMILES CCCCCCCCCCCCCCCCCCCCCCCCCCCCCC

Specifications

Melting Point 66°C
Color White
Boiling Point 200°C
Quantity 5 g
Formula Weight 422.83
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Triacontane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.