Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Triacsin C, Acyl-CoA Synthetase Inhibitor, >90% Purity, 100 μg

Catalog No. 502583340
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-3340 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-3340 Supplier Abcam Supplier No. AB141888100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potent, specific inhibitor of acyl-CoA synthetase that blocks cholesteryl ester and triacylglycerol synthesis.

  • Potent and specific inhibitor of acyl-CoA synthetase
  • Ki values of 15 nM and 2 μM for Faa2p and Faa4p respectively
  • Inhibits the synthesis of cholesteryl ester and triacylglycerol in vitro
  • Orally active vasodilator agent
  • Effective in vivo and in vitro
  • Molecular weight: 207.27 Da
  • Purity: >90%
  • Supplied as a liquid; 100% Triacsin C

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Acyl-CoA Synthetase
Content And Storage Store at -20°C.
Form Liquid
Shipping Condition Ambient
Substrate Properties Chemical Substrate
Quantity 100 μg
For Use With (Application) Acyl-CoA synthetase inhibition, synthesis of cholesteryl ester and triacylglycerol, vasodilation
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.