Learn More
Description
MA3-927 detects triadin from mouse, rabbit and rat tissues. MA3-927 has been successfully used in Western blot, immunohistochemistry, immunoprecipitation and ELISA procedures. By Western blot, this antibody detects a 95 kDa protein representing triadin from rabbit skeletal muscle extracts. Immunohistochemical staining of triadin in rat skeletal muscle with MA3-927 results in striated staining consistent with sarcoplasmic reticulum localization. The MA3-927 antigen is purified rabbit fast skeletal muscle triadin.
Specifications
Specifications
| Antigen | Triadin |
| Applications | ELISA, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | GE 4.90 |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | Ascites with 0.05% sodium azide |
| Gene | TRDN |
| Gene Accession No. | E9Q9K5, Q28820, Q9QX75 |
| Gene Alias | 2310045H21Rik; cardiac triadin; cardiac triadin isoform 1; cardiac triadin isoform 2; cardiac triadin isoform 3; CPVT5; EG432451; junctional sarcoplasmic reticulum glycoprotein; TDN; Trdn; Triadin; triadin 1; triadin 32 kDa (TRISK 32); triadin 49 kDa (TRISK 49); triadin 95; TRISK |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
