Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triallyl Isocyanurate (stabilized with BHT) 96.0+%, TCI America™
SDP

Catalog No. I0279100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0279100G 100 g
I027925G 25 g
I0279500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. I0279100G Supplier TCI America Supplier No. I0279100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1025-15-6
Molecular Formula C12H15N3O3
Molecular Weight (g/mol) 249.27
MDL Number MFCD00006554
InChI Key KOMNUTZXSVSERR-UHFFFAOYSA-N
Synonym triallyl isocyanurate, isocyanuric acid triallyl ester, 1,3,5-triallyl-1,3,5-triazinane-2,4,6-trione, triallylisocyanurate, taic, diak 7, 1,3,5-triallylisocyanurate, 1,3,5-triallyl isocyanurate, 1,3,5-triallylisocyanuric acid, 1,3,5-tri-2-propenyl-1,3,5-triazine-2,4,6 1h,3h,5h-trione
PubChem CID 13931
IUPAC Name 1,3,5-tris(prop-2-enyl)-1,3,5-triazinane-2,4,6-trione
SMILES C=CCN1C(=O)N(C(=O)N(C1=O)CC=C)CC=C

Specifications

Color White-Yellow
Boiling Point 140°C
Quantity 100 g
Formula Weight 249.27
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Triallyl Isocyanurate (stabilized with BHT)
EINECSNumber (5)-1047
RTECSNumber XZ1915000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.