Learn More
YMC America, Inc TRIART C18 1.9UM 100X2.1MM ID
SDP
Supplier:  YMC America, Inc TA12SP910Q1PT
Encompass_Preferred
UHPLC Column, YMC-Triart C18, 1.9um, 120Å, 100x2.1mm ID
Catalog No. 50-109-7851
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.