Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tribromoethylene 97.0+%, TCI America™
SDP

Catalog No. T079425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T079425G 25 g
T0794250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T079425G Supplier TCI America Supplier No. T079425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 598-16-3
Molecular Formula C2HBr3
Molecular Weight (g/mol) 264.74
MDL Number MFCD00039164
InChI Key OZVJKTHTULCNHB-UHFFFAOYSA-N
PubChem CID 11712
IUPAC Name 1,1,2-tribromoethene
SMILES BrC=C(Br)Br

Specifications

Color Yellow
Boiling Point 163°C
Quantity 25 g
Formula Weight 264.74
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Tribromoethylene
RTECSNumber KX4300000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.