Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tribromofluoromethane, 99%, stab. with copper

Catalog No. AAA1812406
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1812406 5 g
AAA1812414 25 g
AAA1812418 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1812406 Supplier Thermo Scientific Chemicals Supplier No. A1812406
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 353-54-8 | CBr3F | 270.721 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 353-54-8
Molecular Formula CBr3F
Molecular Weight (g/mol) 270.721
MDL Number MFCD00000366
InChI Key IHZAEIHJPNTART-UHFFFAOYSA-N
Synonym fluorotribromomethane, methane, tribromofluoro, tribromo fluoro methane, tribromo-fluoro-methane, fluortribrommethan, tribrom fluor methan, cfbr3, fluoro tribromo methane, acmc-209igd, tribromofluoromethane
PubChem CID 67707
IUPAC Name tribromo(fluoro)methane
SMILES C(F)(Br)(Br)Br

Specifications

Melting Point -74°C
Density 2.765
Boiling Point 106°C to 107°C
Flash Point None
Refractive Index 1.5255
Linear Formula FCBr3
Quantity 5 g
Beilstein 1732796
Formula Weight 270.72
Percent Purity 99%
Chemical Name or Material Tribromofluoromethane, stabilized with copper
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 206-535-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.