Learn More
Sigma Organic Chemistry Tributylamine | 10ML | 102-82-9 | MFCD00009431
Supplier: Sigma Organic Chemistry 9078110ML

Tributylamine, 10ML
About This Item:
Linear Formula: [CH3(CH2)3]3N
Molecular Weight: 185.35
CAS Number: 102-82-9
MDL Number: MFCD00009431
UNSPSC Code: 12352100
Purity: >=99.5% (GC)
Boiling Point: 216 C (lit.)
Melting Point: -70 C (lit.)
Density: 0.778 g/mL at 25 C (lit.)
Refractive Index: n20/D 1.428 (lit.); n20/D 1.429
Storage: room temp
EINECS Number: 203-058-7
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.