Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethylacetyl chloride, 99%

Catalog No. AC140050010
Click to view available options
Quantity:
5 g
250 g
1 kg
Packaging:
Glass bottle

Catalog No. AC140050010

Supplier: Thermo Scientific Chemicals 140050010

Only null left
Add to Cart

CAS: 3282-30-2 | C5H9ClO | 120.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3282-30-2
Molecular Formula C5H9ClO
Molecular Weight (g/mol) 120.58
MDL Number MFCD00000709
InChI Key JVSFQJZRHXAUGT-UHFFFAOYSA-N
Synonym pivaloyl chloride, trimethylacetyl chloride, propanoyl chloride, 2,2-dimethyl, pivalyl chloride, pivaloylchloride, 2,2-dimethylpropionyl chloride, pivalic acid chloride, pivalolyl chloride, unii-jq82j0o21t, 2,2-dimethylpropionylchloride
PubChem CID 62493
IUPAC Name 2,2-dimethylpropanoyl chloride
SMILES CC(C)(C)C(Cl)=O

Specifications

Melting Point -56.0°C
Color Colorless to Pink
Density 0.9800g/mL
Boiling Point 105.0°C
Flash Point 14°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.4110 to 1.4130
Linear Formula (CH3)3CCOCl
Quantity 1 kg
Beilstein 02, 320
Fieser 01,1229; 08,404; 11,489
Specific Gravity 0.98
Solubility Information Solubility in water: hydrolysis., miscible with most common aprotic organic solvents
Viscosity 0.86 mPa.s (20°C)
Formula Weight 120.58
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Trimethylacetyl chloride, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Fatal if inhaled.
Causes severe skin burns and eye damage.
May be corrosive to metals.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 221-921-6
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.