Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tributylsilane 98.0+%, TCI America™
SDP

Catalog No. T204325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T204325G 25 g
T20435G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T204325G Supplier TCI America Supplier No. T204325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 998-41-4
Molecular Formula C12H27Si
Molecular Weight (g/mol) 199.43
MDL Number MFCD00009415
InChI Key ISEIIPDWJVGTQS-UHFFFAOYSA-N
PubChem CID 6327711
IUPAC Name tributylsilyl
SMILES CCCC[Si](CCCC)CCCC

Specifications

Color Colorless
Boiling Point 103°C
Quantity 25 g
Formula Weight 200.44
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Tributylsilane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.