Learn More
Medchemexpress LLC Tricarbonylchloro(glycinato)ruthenium (II) | 475473-26-8 | 97.5% | 294.61 g·mol⁻¹ | C5H4ClNO5Ru | 100 MG
SDP
Supplier:  Medchemexpress LLC HY100581100MG
Encompass_Preferred
CORM-3 is a ruthenium-based carbon monoxide (CO) releasing molecule used as a research reagent to deliver CO in biological studies. It is used to investigate CO-mediated signaling, oxidative stress, and inflammatory responses in cells and tissues.
  • Releases carbon monoxide for biological research.
  • Attenuates NF-κB p65 nuclear translocation and reduces reactive oxygen species.
  • Molecular formula C5H4ClNO5Ru; molecular weight 294.61 g·mol⁻¹.
  • Reported purity approximately 97.5%.
  • Available in multiple package sizes, including 100 mg.
Catalog No. 50-002-12754