Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trichloroacetic Acid, 100% (w/v) Aqueous Solution, Spectrum™ Chemical
SDP

Catalog No. 18607543 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-543 500 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-607-543 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. T184500MLGL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

T-184, 76-03-9

Spectrum™ Chemical Trichloroacetic Acid, 100 Percent (w/v) Aqueous Solution , also known as TCA, is used in biochemistry to help the precipitation of macromolecules. It is also used in the cosmetic industry as a chemical peel and also as a wart remover.

Chemical Identifiers

CAS 76-03-9
Molecular Formula C2HCl3O2
Molecular Weight (g/mol) 163.38
InChI Key YNJBWRMUSHSURL-UHFFFAOYSA-N
IUPAC Name trichloroacetic acid
SMILES OC(=O)C(Cl)(Cl)Cl

Specifications

CAS Max % 1
Quantity 500 mL
Packaging Amber Glass Bottle
Percent Purity 100% ±2%
Grade Chemical Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.