Learn More
Sigma Organic Chemistry Trichloroacetic acid | 1KG | 76-03-9 | MFCD00004177
Supplier: Sigma Organic Chemistry T48851KG

Trichloroacetic acid, 1KG
About This Item:
Synonym(s): TCA
Linear Formula: Cl3CCOOH
Molecular Weight: 163.39
CAS Number: 76-03-9
MDL Number: MFCD00004177
UNSPSC Code: 12352100
Purity: >=99.0% (titration)
Boiling Point: 196 C (lit.)
Melting Point: 54-58 C (lit.)
Density: 1.62 g/mL at 25 C (lit.)
Refractive Index: n20/D 1.62 (lit.)
Storage: 2-8C
EINECS Number: 200-927-2
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.