Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trichloroacetic Acid, 5% (w/v), Ricca Chemical
SDP

Catalog No. 868016 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
500 mL
1 L
4 L
20 L
Packaging:
Amber Glass Bottle
Cubitainer
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
8680-16 500 mL Amber Glass Bottle
8680-4 120 mL Amber Glass Bottle
8680-32 1 L Amber Glass Bottle
R86800004C 4 L Amber Glass Bottle
R868000020F 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 8680-16 Supplier Ricca Chemical Company Supplier No. 868016
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7732-18-5 , 76-03-9
Molecular Formula C2HCl3O2
Molecular Weight (g/mol) Mixture
InChI Key YNJBWRMUSHSURL-UHFFFAOYSA-N
Synonym trichloroacetic acid, trichloroethanoic acid, aceto-caustin, trichloroacetic acid, acetic acid, trichloro, trichloressigsaeure, konesta, acide trichloracetique, acido tricloroacetico, amchem grass killer
PubChem CID 6421
ChEBI CHEBI:30956
IUPAC Name trichloroacetic acid
SMILES OC(=O)C(Cl)(Cl)Cl

Specifications

Percent Purity 5% w/v
Grade Laboratory
Concentration or Composition (by Analyte or Components) 5% (w/v) Aqueous Solution
Quantity 500 mL
Packaging Amber Glass Bottle
Chemical Name or Material Trichloroacetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.