Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trichloroacetic Acid, Crystal, Reagent, ACS, 99%, Spectrum™ Chemical
SDP

Catalog No. 18607552 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
6 x 125 g
6 x 500 g
25 kg
Packaging:
Amber Glass Bottle
Poly Pail
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-552 25 g Amber Glass Bottle
18-607-553 125 g Amber Glass Bottle
18-607-555 500 g Amber Glass Bottle
18-607-554 6 x 125 g Amber Glass Bottle
18-607-557 2.5 kg Amber Glass Bottle
18-607-556 6 x 500 g Amber Glass Bottle
18-607-558 25 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18-607-552 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. T110525GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

T1105, 76-03-9, C2HCl3O2

Spectrum™ Chemical Trichloroacetic Acid, Crystal, Reagent, ACS , also known as TCA, is used in biochemistry to help the precipitation of macromolecules. It is also used in the cosmetic industry as a chemical peel and also as a wart remover. As an ACS grade Reagent, Spectrum Chemical manufactured trichloroacetic acid is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness.

Chemical Identifiers

CAS 76-03-9
Molecular Formula C2HCl3O2
Molecular Weight (g/mol) 163.38
InChI Key YNJBWRMUSHSURL-UHFFFAOYSA-N
IUPAC Name trichloroacetic acid
SMILES OC(=O)C(Cl)(Cl)Cl

Specifications

CAS Max % 1
Percent Purity 99%
Grade ACS
Quantity 25 g
Packaging Amber Glass Bottle
Residue after Ignition 0.0003
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.