Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trichloroacetic Acid (Flakes or Crystals), Fisher BioReagents

Catalog No. BP555500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
250 g
500 g
1 kg
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP555-500 500 g Glass Bottle
BP555-250 250 g Glass Bottle
BP555-1 1 kg Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. BP555-500

missing translation for 'validMainframeMsg'

TCA is used to precipitate proteins and nucleic acids from solution.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76-03-9
Molecular Formula C2HCl3O2
Molecular Weight (g/mol) 163.378
InChI Key YNJBWRMUSHSURL-UHFFFAOYSA-N
Synonym trichloroacetic acid, trichloroethanoic acid, aceto-caustin, trichloroacetic acid, acetic acid, trichloro, trichloressigsaeure, konesta, acide trichloracetique, acido tricloroacetico, amchem grass killer
PubChem CID 6421
ChEBI CHEBI:30956
IUPAC Name 2,2,2-trichloroacetic acid
SMILES C(=O)(C(Cl)(Cl)Cl)O

Specifications

pH 1.2
Melting Point 52°C
Boiling Point 196°C
Color White
Assay Percent Range ≥99.0 %
Linear Formula CCl3COOH
UN Number 1839
Merck Index 15, 9792
Formula Weight 163.39
Percent Purity ≥99.0%
Density 1.620g/cm³
Quantity 500 g
Ignition Residue 0.030% max.
Packaging Glass Bottle
Physical Form Solid
Vapor Pressure 1.2mbar at 50°C, 0.08mbar at 25°C
Chemical Name or Material Trichloroacetic Acid
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Possible cancer hazard. May cause cancer based on animal data. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Hygroscopic. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required.

NFPA

Health:3
Flammability:1
Instability:1
DOTInformation DOT Class 8, : Corrosive
Recommended Storage RT
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.