Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trichloroacetic Acid, MP Biomedicals™

Catalog No. MP215259290 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215259290 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP215259290 Supplier MP Biomedicals, Inc Supplier No. 0215259290
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used as a decalcifier and fixative in microscopy. Trichloroacetic Acid, MP Biomedicals is used in protein precipitation and has been used to determine protein concentration by quantitative precipitation. A protocol for the precipitation of nucleic acids c

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76-03-9
Molecular Formula C2HCl3O2
Molecular Weight (g/mol) 163.378
InChI Key YNJBWRMUSHSURL-UHFFFAOYSA-N
Synonym trichloroacetic acid, trichloroethanoic acid, aceto-caustin, trichloroacetic acid, acetic acid, trichloro, trichloressigsaeure, konesta, acide trichloracetique, acido tricloroacetico, amchem grass killer
PubChem CID 6421
ChEBI CHEBI:30956
IUPAC Name 2,2,2-trichloroacetic acid
SMILES C(=O)(C(Cl)(Cl)Cl)O

Specifications

Solubility Information Soluble in water; very soluble in ethanol or ether. Decomposed by heating with caustic alkalies into chloroform and alkali carbonate.
Formula Weight 163.4
Percent Purity ≥99.0%
Grade ACS
Quantity 500 g
Physical Form Powder
Recommended Storage Room Temperature (15–30°C). Store Desiccated.
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.