Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trichloroacetic Acid Solution, MP Biomedicals™

Catalog No. MP219605791 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219605791 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP219605791 Supplier MP Biomedicals, Inc Supplier No. 0219605791
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Commonly used to precipitate proteins, DNA and RNA. Trichloroacetic Acid Solution, MP Biomedicals is an acetic acid analog that has also found use as a decalcifier and fixative in microscopy, in organic synthesis and for protein sequencing and detecting a

In the presence of SDS, Trichloroacetic acid will precipitate proteins which can then be quantified by the Lowry method. Trichloroacetic acid has also been used to determine protein concentration by quantitative precipitation. It is also used as a decalcifier and fixative in microscopy. A protocol exists for the precipitation of nucleic acids.

Rats exposed to chronic sublethal amounts of trichloroacetic acid displayed an increase in serum bilirubin with a decrease in hematological proteins and cholesterols along with significant decreases in red blood corpuscles, mean cell volume, mean corpuscular hemoglobin, hemoglobin and hematocrit. In mice the production of liver tumors was associated to Trichloroacetic acid, which is also a known mouse hepatocarinogen. On the guinea pig trichloroacetic acid was observed to be a mild alergen on the skin.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76-03-9
Molecular Formula C2HCl3O2
Molecular Weight (g/mol) 163.378
InChI Key YNJBWRMUSHSURL-UHFFFAOYSA-N
Synonym trichloroacetic acid, trichloroethanoic acid, aceto-caustin, trichloroacetic acid, acetic acid, trichloro, trichloressigsaeure, konesta, acide trichloracetique, acido tricloroacetico, amchem grass killer
PubChem CID 6421
ChEBI CHEBI:30956
IUPAC Name 2,2,2-trichloroacetic acid
SMILES C(=O)(C(Cl)(Cl)Cl)O

Specifications

Formula Weight 163.4g/mol
Quantity 1 g
Physical Form Solution
Recommended Storage Store at 4°C.
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.