Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trichloroacetyl Isocyanate 97.0+%, TCI America™
SDP

Catalog No. T104725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T104725G 25 g
T10471G 1 g
T10475G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T104725G Supplier TCI America Supplier No. T104725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3019-71-4
Molecular Formula C3Cl3NO2
Molecular Weight (g/mol) 188.39
MDL Number MFCD00002033
InChI Key GRNOZCCBOFGDCL-UHFFFAOYSA-N
Synonym trichloroacetyl isocyanate, acetyl isocyanate, trichloro, trichloroacetylisocyanate, isocyanic acid trichloroacetyl ester, unii-qxv1p5j6jl, qxv1p5j6jl, acetyl isocyanate, 2,2,2-trichloro, 2,2,2-trichloroethanecarbonyl isocyanate, trichloroethanecarbonyl isocyanate, trichloroacetyl isocyanate, nmr grade
PubChem CID 76400
IUPAC Name trichloroethanecarbonyl isocyanate
SMILES ClC(Cl)(Cl)C(=O)N=C=O

Specifications

Color Yellow
Boiling Point 85°C
Quantity 25 g
UN Number 2206
Formula Weight 188.39
Percent Purity ≥97.0% (N)
Physical Form Liquid
Chemical Name or Material Trichloroacetyl Isocyanate
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.