Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trichloroethylene, ≥99.5%, ACS Reagent, Solstice

Catalog No. 60045683 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-045-683 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 60-045-683 Supplier Solstice Supplier No. 2514021L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

ACS Reagent,≥99.5%

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-01-6
Molecular Formula C2HCl3
Molecular Weight (g/mol) 131.38
MDL Number MFCD00000838
InChI Key XSTXAVWGXDQKEL-UHFFFAOYSA-N
Synonym trichloroethylene, trichloroethene, ethene, trichloro, trilene, trichlorethylene, ethinyl trichloride, ethylene trichloride, acetylene trichloride, chlorilen, narcogen
PubChem CID 6575
ChEBI CHEBI:16602
IUPAC Name 1,1,2-trichloroethene
SMILES C(=C(Cl)Cl)Cl

Specifications

Melting Point -84.8°C
Density 1.463g/mL at 25°C
Boiling Point 86.7°C
Ignition Point 770°F
Packaging Glass Bottle
Linear Formula ClCH=CCl2
Quantity 1 L
UN Number UN1710
Beilstein 1736782
Vapor Density 4.5 (vs air)
Vapor Pressure 61mmHg (20°C)
Formula Weight 131.39g/mol
Percent Purity ≥99.5%
Grade ACS Reagent
Physical Form Liquid
Chemical Name or Material Trichloroethylene
Show More Show Less
Hazard Statement P201-P261-P273-P281-P305 + P351 + P338-P308 + P313
Precautionary Statement H315-H319-H336-H341-H350-H412
DOTInformation 6.1/PGIII
EINECSNumber 201-167-4
RTECSNumber KX4550000
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.