CAS RN 79-01-6
Trichloroethylene, ACS, 99.5% min
CAS: 79-01-6 Molecular Formula: C2HCl3 Molecular Weight (g/mol): 131.38 MDL Number: MFCD00000838 InChI Key: XSTXAVWGXDQKEL-UHFFFAOYSA-N Synonym: trichloroethylene,trichloroethene,ethene, trichloro,trilene,trichlorethylene,ethinyl trichloride,ethylene trichloride,acetylene trichloride,chlorilen,narcogen PubChem CID: 6575 ChEBI: CHEBI:16602 IUPAC Name: 1,1,2-trichloroethene SMILES: C(=C(Cl)Cl)Cl
Trichloroethylene, 99+%, extra pure, stabilized
CAS: 79-01-6 MDL Number: MFCD00000838 InChI Key: XSTXAVWGXDQKEL-UHFFFAOYSA-N Synonym: trichloroethylene,trichloroethene,ethene, trichloro,trilene,trichlorethylene,ethinyl trichloride,ethylene trichloride,acetylene trichloride,chlorilen,narcogen PubChem CID: 6575 ChEBI: CHEBI:16602 IUPAC Name: 1,1,2-trichloroethene SMILES: C(=C(Cl)Cl)Cl
Trichloroethylene, 98%
CAS: 79-01-6 Molecular Formula: C2HCl3 Molecular Weight (g/mol): 131.38 MDL Number: MFCD00000838 InChI Key: XSTXAVWGXDQKEL-UHFFFAOYSA-N Synonym: trichloroethylene,trichloroethene,ethene, trichloro,trilene,trichlorethylene,ethinyl trichloride,ethylene trichloride,acetylene trichloride,chlorilen,narcogen PubChem CID: 6575 ChEBI: CHEBI:16602 IUPAC Name: 1,1,2-trichloroethene SMILES: C(=C(Cl)Cl)Cl
Trichloroethylene, 99.6%, ACS reagent, stabilized
CAS: 79-01-6 Molecular Formula: C2HCl3 Molecular Weight (g/mol): 131.39 MDL Number: MFCD00000838 InChI Key: XSTXAVWGXDQKEL-UHFFFAOYSA-N Synonym: trichloroethylene,trichloroethene,ethene, trichloro,trilene,trichlorethylene,ethinyl trichloride,ethylene trichloride,acetylene trichloride,chlorilen,narcogen PubChem CID: 6575 ChEBI: CHEBI:16602 IUPAC Name: 1,1,2-trichloroethene SMILES: C(=C(Cl)Cl)Cl
Trichloroethylene, ACS, 99.5%, Spectrum™ Chemical
CAS: 79-01-6 Molecular Formula: C2HCl3 Molecular Weight (g/mol): 131.38 InChI Key: XSTXAVWGXDQKEL-UHFFFAOYSA-N IUPAC Name: 1,1,2-trichloroethene SMILES: ClC=C(Cl)Cl
Trichloroethylene, ACS Reagent (Trichloroethene), Reagents
CAS: 79-01-6 Molecular Formula: C2HCl3 Molecular Weight (g/mol): 131.38 InChI Key: XSTXAVWGXDQKEL-UHFFFAOYSA-N Synonym: Trichloroethene, 1,1,2-Trichloroethene IUPAC Name: 1,1,2-trichloroethene SMILES: ClC=C(Cl)Cl
Trichloroethylene, Purified (Trichloroethene), Reagents
CAS: 79-01-6 Molecular Formula: C2HCl3 Molecular Weight (g/mol): 131.38 InChI Key: XSTXAVWGXDQKEL-UHFFFAOYSA-N Synonym: Trichloroethene, 1,1,2-Trichloroethene IUPAC Name: 1,1,2-trichloroethene SMILES: ClC=C(Cl)Cl
Trichloroethylene, ≥99.5%, ACS Reagent, Solstice
CAS: 79-01-6 Molecular Formula: C2HCl3 Molecular Weight (g/mol): 131.38 MDL Number: MFCD00000838 InChI Key: XSTXAVWGXDQKEL-UHFFFAOYSA-N Synonym: trichloroethylene,trichloroethene,ethene, trichloro,trilene,trichlorethylene,ethinyl trichloride,ethylene trichloride,acetylene trichloride,chlorilen,narcogen PubChem CID: 6575 ChEBI: CHEBI:16602 IUPAC Name: 1,1,2-trichloroethene SMILES: C(=C(Cl)Cl)Cl
Avantor J.T.Baker Trichloroethylene, Stabilized, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
CAS: 79-01-6 Molecular Formula: C2HCl3 Molecular Weight (g/mol): 131.38 InChI Key: XSTXAVWGXDQKEL-UHFFFAOYSA-N Synonym: trichloroethylene,trichloroethene,ethene, trichloro,trilene,trichlorethylene,ethinyl trichloride,ethylene trichloride,acetylene trichloride,chlorilen,narcogen PubChem CID: 6575 ChEBI: CHEBI:16602 IUPAC Name: 1,1,2-trichloroethene SMILES: C(=C(Cl)Cl)Cl
Trichloroethylene, anhydrous, >99%, Thermo Scientific™
CAS: 79-01-6 Molecular Formula: C2HCl3 Molecular Weight (g/mol): 131.38 MDL Number: MFCD00000838 InChI Key: XSTXAVWGXDQKEL-UHFFFAOYSA-N Synonym: trichloroethylene,trichloroethene,ethene, trichloro,trilene,trichlorethylene,ethinyl trichloride,ethylene trichloride,acetylene trichloride,chlorilen,narcogen PubChem CID: 6575 ChEBI: CHEBI:16602 IUPAC Name: 1,1,2-trichloroethene SMILES: C(=C(Cl)Cl)Cl
Trichloroethene in Dimethyl Sulfoxide, SPEX CertiPrep™
CAS: 79-01-6 Molecular Formula: C2HCl3 Molecular Weight (g/mol): 131.38 InChI Key: XSTXAVWGXDQKEL-UHFFFAOYSA-N PubChem CID: 6575 ChEBI: CHEBI:16602 IUPAC Name: 1,1,2-trichloroethene SMILES: C(=C(Cl)Cl)Cl
Trichloroethene, SPEX CertiPrep™
CAS: 79-01-6 Molecular Formula: C2HCl3 Molecular Weight (g/mol): 131.38 InChI Key: XSTXAVWGXDQKEL-UHFFFAOYSA-N PubChem CID: 6575 ChEBI: CHEBI:16602 IUPAC Name: 1,1,2-trichloroethene SMILES: C(=C(Cl)Cl)Cl