Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trichloroethylene, ACS Reagent (Trichloroethene), Reagents

Catalog No. 01337639
Change view
Click to view available options
Quantity:
500 mL
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-639 4 L
01-337-638 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-337-639 Supplier Reagents Holdings Llc Supplier No. C13420004C

missing translation for 'validMainframeMsg'

Trichloroethylene, ACS Reagent (Trichloroethene), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 79-01-6
Molecular Formula C2HCl3
Molecular Weight (g/mol) 131.38
InChI Key XSTXAVWGXDQKEL-UHFFFAOYSA-N
Synonym Trichloroethene, 1,1,2-Trichloroethene
IUPAC Name 1,1,2-trichloroethene
SMILES ClC=C(Cl)Cl

Specifications

Boiling Range 87°C
Melting Point -84.8°C
Color Colorless
Boiling Point 87°C
Melting Range -84.8°C
Packaging Glass Bottle
Quantity 4 L
Specific Gravity 1.47
Grade ACS
Physical Form Liquid
Chemical Name or Material Trichloroethylene, ACS Reagent (Trichloroethene)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.