Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trichlorovinylsilane 98.0+%, TCI America™
SDP

Catalog No. T040725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T040725G 25 g
T0407100G 100 g
T0407500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T040725G Supplier TCI America Supplier No. T040725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 75-94-5
Molecular Formula C2H3Cl3Si
Molecular Weight (g/mol) 161.481
MDL Number MFCD00000482
InChI Key GQIUQDDJKHLHTB-UHFFFAOYSA-N
Synonym trichlorovinylsilane, vinyltrichlorosilane, trichloro vinyl silane, vtcs, silane, trichloroethenyl, silane, trichlorovinyl, trichlorovinylsilicon, vinyl trichlorosilane, trichlorovinyl silicane, vinylsilicon trichloride
PubChem CID 6413
IUPAC Name trichloro(ethenyl)silane
SMILES C=C[Si](Cl)(Cl)Cl

Specifications

Color Yellow
Boiling Point 92°C
Quantity 25 g
UN Number 1305
Formula Weight 161.48
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Trichlorovinylsilane
EINECSNumber (2)-2037
RTECSNumber VV6125000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.