Learn More
Cayman Chemical TrIchoderma VIrIdIol 1mg
SDP
Supplier:  Cayman Chemical 196071 MG
Encompass_Preferred
An aminothiazole that acts as a SIRT-rearranging ligand to selectively inhibit SIRT2 (IC50 140 nM) without effect on SIRT1 or SIRT3-6 increases a-tubulin acetylation in HeLa cells
Catalog No. 50-298-2460
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.