Learn More
Thermo Scientific™ Trichophyton Agar #6
Description
Differentiate and identify Trichophyton species based on nutritional requirements with ready-to-use Trichophyton Agar No. 6.
Differentiate and identify Trichophyton species based on nutritional requirements using Thermo Scientific™ Remel™ Trichophyton Agar No. 6 (Ammonium Nitrate Agar Base). Different Trichophyton species resemble one another closely in culture and are difficult to differentiate on the basis of colony formation or microscopic morphology alone. Georg and Camp developed Trichophyton Agars for differentiation and identification of Trichophyton isolates based on specific vitamin and amino acid requirements1.
Trichophyton Agars are to be used with Trichophyton isolates that have been presumptively identified by gross colony characteristics and microscopic morphology 2-5.
Trichophyton Agar No. 6 (Ammonium nitrate agar base) contains
Specifications
Specifications
| Format | 20mm x 113mm Tube - Slant |
| Media Type | Trichophyton Agar No. 3 |
| Product Type | Agar Slants |
| Quantity | 20/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.