Learn More
ATCC Trichosporon inkin (Strain B-4475) - Frozen Yeast Culture, ATCC 66343
Supplier: ATCC 66343
Trichosporon inkin is a clinical yeast strain provided as a frozen ampoule containing viable cells suspended in cryoprotectant for use in biomedical research. The culture (strain designation B-4475) should be handled under BSL-2 conditions and recovered according to supplier thawing and inoculation instructions.
- Supplied as a frozen ampoule containing viable cells in cryoprotectant.
- Strain designation: B-4475.
- Storage: keep at -80°C or colder; do not store at -20°C.
- Recommended medium: Emmons' modification of Sabouraud's agar/broth (ATCC Medium 28).
- Incubation temperature: 24°C; inspect for growth for up to 4 weeks.
- Isolation source: human lung from a patient with X-linked chronic granulomatous disease.
- Biosafety: handle at biosafety level 2 (BSL-2).
- Intended use: biomedical research and development; follow established aseptic techniques for recovery.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.